Assay Details
Gene Symbol: Sh2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207605.3 | NP_997488.2 | 1 - 2 | 66 | 67 | |
GenBank mRNA | AF468787.2 | - | 1 - 2 | 58 | 67 | |
BC088087.1 | - | 1 - 2 | 66 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
