Assay Details
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002016.2 | NP_001002016.2 | 11 - 12 | 2173 | 80 | |
GenBank mRNA | X66870.1 | - | 11 - 12 | 2131 | 80 | |
X76297.1 | - | 11 - 12 | 1895 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
nuclear envelope organization
spermatogenesis
muscle organ development
regulation of cell migration
establishment or maintenance of microtubule cytoskeleton polarity
protein localization to nucleus
ventricular cardiac muscle cell development
cellular response to hypoxia
negative regulation of release of cytochrome c from mitochondria
positive regulation of cell aging
regulation of protein localization to nucleus
negative regulation of extrinsic apoptotic signaling pathway
structural molecule activity
nucleus organization
nuclear envelope organization
spermatogenesis
muscle organ development
regulation of cell migration
establishment or maintenance of microtubule cytoskeleton polarity
protein localization to nucleus
ventricular cardiac muscle cell development
cellular response to hypoxia
negative regulation of release of cytochrome c from mitochondria
positive regulation of cell aging
regulation of protein localization to nucleus
negative regulation of extrinsic apoptotic signaling pathway
structural molecule activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P097654 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P013725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80863 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P097654 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P013725 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P097654 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P013725 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P013725 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P097654 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P416761 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89626 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35429 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10824330 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824326 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824332 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824329 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824328 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824325 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824336 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824338 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824323 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824324 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824333 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824335 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824337 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824334 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824331 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824327 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10824322 | - | - |
Rat Genome 230 2.0 Array | 1368055_a_at | - | - |
Rat Genome 230 2.0 Array | 1368054_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368054_at | - | - |
Rat Genome 230 2.0 Array | 1368055_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368054_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368055_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368054_at | - | - |
Affymetrix HT Rat Focus Array | 1368055_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200027337.rn.1 | TC0200004038.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200027335.rn.1 | TC0200004038.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200027336.rn.1 | TC0200004038.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200027338.rn.1 | TC0200004038.rn.1 | RTA TC |
Rat Expression Set 230 | 1368054_PM_at | - | - |
Rat Expression Set 230 | 1368055_PM_a_at | - | - |
Rat Expression Set 230 | 1368055_a_at | - | - |
Rat Expression Set 230 | 1368054_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10824322 | - | - |
Rat Genome U34 Set | X66870_at | - | - |
Rat Genome U34 Set | rc_AI013260_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21748338 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374095 | - | - |
RatRef-12 Expression BeadChip | ILMN_61746 | - | - |
