Assay Details
Gene Symbol: Ntrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021589.1 | NP_067600.1 | 4 - 5 | 486 | 69 | |
XM_006232746.2 | XP_006232808.1 | 4 - 5 | 477 | 69 | ||
GenBank mRNA | AF174586.1 | - | 1 - 2 | 80 | 69 | |
M85214.1 | - | 4 - 5 | 486 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
