Assay Details
Gene Symbol: Tdo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022403.2 | NP_071798.1 | 2 - 3 | 184 | 104 | |
GenBank mRNA | BC089802.1 | - | 2 - 3 | 180 | 104 | |
M55167.1 | - | 2 - 3 | 184 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
