Assay Details
Gene Symbol: Dusp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106624.2 | NP_001100094.1 | 3 - 4 | 538 | 61 | |
XM_008763374.1 | XP_008761596.1 | 4 - 5 | 1114 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
