Assay Details
Gene Symbol: Mgst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134349.3 | NP_599176.1 | 1 - 2 | 40 | 65 | |
GenBank mRNA | BC063150.1 | - | 1 - 2 | 38 | 65 | |
BP494884.1 | - | 1 - 2 | 39 | 65 | ||
CB711033.1 | - | 1 - 2 | 56 | 65 | ||
FQ209427.1 | - | 1 - 2 | 61 | 65 | ||
FQ209464.1 | - | 1 - 2 | 53 | 65 | ||
FQ209465.1 | - | 1 - 2 | 60 | 65 | ||
FQ211049.1 | - | 1 - 2 | 60 | 65 | ||
FQ212841.1 | - | 1 - 2 | 61 | 65 | ||
FQ218096.1 | - | 1 - 2 | 43 | 65 | ||
FQ218830.1 | - | 1 - 2 | 68 | 65 | ||
FQ219010.1 | - | 1 - 2 | 62 | 65 | ||
FQ219410.1 | - | 1 - 2 | 34 | 65 | ||
FQ219464.1 | - | 1 - 2 | 66 | 65 | ||
FQ219581.1 | - | 1 - 2 | 62 | 65 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC063150.1, NM_134349.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
