Assay Details
Gene Symbol: Fkbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191863.1 | NP_001178792.1 | 5 - 6 | 834 | 91 | |
GenBank mRNA | AF531427.1 | - | 4 - 5 | 501 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
