Assay Details
Gene Symbol: Arhgdib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009600.1 | NP_001009600.1 | 2 - 3 | 246 | 90 | |
XM_006237561.2 | XP_006237623.1 | 2 - 3 | 270 | 90 | ||
XM_006237562.2 | XP_006237624.1 | 2 - 3 | 401 | 90 | ||
XM_006237563.2 | XP_006237625.1 | 3 - 4 | 332 | 90 | ||
XM_006237564.2 | XP_006237626.1 | 2 - 3 | 361 | 90 | ||
XM_006237565.2 | XP_006237627.1 | 2 - 3 | 546 | 90 | ||
GenBank mRNA | BC088209.1 | - | 2 - 3 | 246 | 90 | |
FQ225418.1 | - | 2 - 3 | 220 | 90 | ||
FQ233135.1 | - | 2 - 3 | 227 | 90 | ||
FQ234136.1 | - | 2 - 3 | 241 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
