Assay Details
Gene Symbol: Arhgdib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009600.1 | NP_001009600.1 | 3 - 4 | 332 | 158 | |
XM_006237561.2 | XP_006237623.1 | 3 - 4 | 356 | 158 | ||
XM_006237562.2 | XP_006237624.1 | 3 - 4 | 487 | 158 | ||
XM_006237563.2 | XP_006237625.1 | 4 - 5 | 418 | 158 | ||
XM_006237564.2 | XP_006237626.1 | 3 - 4 | 447 | 158 | ||
XM_006237565.2 | XP_006237627.1 | 3 - 4 | 632 | 158 | ||
GenBank mRNA | BC088209.1 | - | 3 - 4 | 332 | 158 | |
FQ225418.1 | - | 3 - 4 | 306 | 158 | ||
FQ233135.1 | - | 3 - 4 | 313 | 158 | ||
FQ234136.1 | - | 3 - 4 | 327 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
