Assay Details
Gene Symbol: Arhgdib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009600.1 | NP_001009600.1 | 1 - 2 | 65 | 81 | |
XM_006237563.2 | XP_006237625.1 | 2 - 3 | 151 | 81 | ||
GenBank mRNA | BC088209.1 | - | 1 - 2 | 65 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC088209.1, NM_001009600.1, XM_006237563.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
