Assay Details
Gene Symbol: Gnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021858.3 | NP_068630.1 | 1 - 2 | 79 | 130 | |
XM_006237383.1 | XP_006237445.1 | 1 - 2 | 1061 | 130 | ||
GenBank mRNA | BC086422.1 | - | 1 - 2 | 82 | 130 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC086422.1, NM_021858.3, XM_006237383.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
