Assay Details
Gene Symbol: Tead4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237424.2 | XP_006237486.1 | 2 - 3 | 2267 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
