Assay Details
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247180.2 | XP_006247242.1 | 6 - 7 | 1440 | 89 | |
XM_006247181.2 | XP_006247243.1 | 7 - 8 | 747 | 89 | ||
XM_006247182.2 | XP_006247244.1 | 6 - 7 | 696 | 89 | ||
XM_008767968.1 | XP_008766190.1 | 6 - 7 | 792 | 89 | ||
XM_008767969.1 | XP_008766191.1 | 7 - 8 | 714 | 89 | ||
GenBank mRNA | BC091435.1 | - | 3 - 4 | 362 | 89 | |
FQ217100.1 | - | 7 - 8 | 944 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
