Assay Details
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100496.1 | NP_001093966.1 | 7 - 8 | 872 | 136 | |
XM_006247180.2 | XP_006247242.1 | 8 - 9 | 1676 | 136 | ||
XM_006247181.2 | XP_006247243.1 | 9 - 10 | 983 | 136 | ||
XM_006247182.2 | XP_006247244.1 | 8 - 9 | 932 | 136 | ||
XM_008767968.1 | XP_008766190.1 | 8 - 9 | 1028 | 136 | ||
XM_008767969.1 | XP_008766191.1 | 9 - 10 | 950 | 136 | ||
GenBank mRNA | BC091435.1 | - | 5 - 6 | 598 | 136 | |
BC166743.1 | - | 7 - 8 | 872 | 136 | ||
FQ217100.1 | - | 9 - 10 | 1180 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
