Assay Details
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100496.1 | NP_001093966.1 | 9 - 10 | 1144 | 87 | |
XM_006247180.2 | XP_006247242.1 | 10 - 11 | 1948 | 87 | ||
XM_006247181.2 | XP_006247243.1 | 11 - 12 | 1255 | 87 | ||
XM_006247182.2 | XP_006247244.1 | 10 - 11 | 1204 | 87 | ||
XM_008767968.1 | XP_008766190.1 | 10 - 11 | 1300 | 87 | ||
XM_008767969.1 | XP_008766191.1 | 11 - 12 | 1222 | 87 | ||
GenBank mRNA | BC091435.1 | - | 7 - 8 | 870 | 87 | |
BC166743.1 | - | 9 - 10 | 1144 | 87 | ||
FQ217100.1 | - | 11 - 12 | 1452 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
ubiquitin-dependent protein catabolic process
glucose homeostasis
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of DNA-binding transcription factor activity
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of type B pancreatic cell apoptotic process
ubiquitin protein ligase binding
negative regulation of transcription by RNA polymerase II
ubiquitin-dependent protein catabolic process
glucose homeostasis
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of DNA-binding transcription factor activity
positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of type B pancreatic cell apoptotic process
ubiquitin protein ligase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P179719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126560 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94410 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P179719 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P179719 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P179719 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P179719 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P187827 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8507 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10737619 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10737611 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10737612 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10737617 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10737613 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10737610 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10737614 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10737618 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10737615 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10737616 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10737609 | - | - |
Rat Genome 230 2.0 Array | 1398367_at | - | - |
Rat Genome 230 2.0 Array | 1398367_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398367_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398367_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000013192.rn.1 | TC1000001471.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000013193.rn.1 | TC1000001471.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000013194.rn.1 | TC1000001471.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000013191.rn.1 | TC1000001471.rn.1 | RTA TC |
Rat Expression Set 230 | 1398367_at | - | - |
Rat Expression Set 230 | 1398367_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10737609 | - | - |
Rat Genome U34 Set | rc_AA799552_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21079308 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353917 | - | - |
