Assay Details
Gene Symbol: Timp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021989.2 | NP_068824.1 | 1 - 2 | 252 | 67 | |
GenBank mRNA | AJ409332.1 | - | 1 - 2 | 252 | 67 | |
BC084714.1 | - | 1 - 2 | 344 | 67 | ||
L31884.1 | - | 1 - 2 | 335 | 67 | ||
S72594.1 | - | 1 - 2 | 356 | 67 | ||
S82718.1 | - | 1 - 2 | 345 | 67 | ||
U14526.1 | - | 1 - 2 | 335 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
