Assay Details
Gene Symbol: Timp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021989.2 | NP_068824.1 | 4 - 5 | 596 | 75 | |
GenBank mRNA | AJ409332.1 | - | 4 - 5 | 596 | 75 | |
BC084714.1 | - | 4 - 5 | 688 | 75 | ||
L31884.1 | - | 4 - 5 | 679 | 75 | ||
S72594.1 | - | 4 - 5 | 700 | 75 | ||
S82718.1 | - | 4 - 5 | 689 | 75 | ||
U14526.1 | - | 4 - 5 | 679 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
spermatogenesis
central nervous system development
aging
negative regulation of cell population proliferation
response to xenobiotic stimulus
response to hormone
negative regulation of endopeptidase activity
regulation of Rap protein signal transduction
response to cytokine
positive regulation of catalytic activity
negative regulation of catalytic activity
regulation of MAPK cascade
positive regulation of MAPK cascade
regulation of neuron differentiation
positive regulation of neuron differentiation
positive regulation of adenylate cyclase activity
negative regulation of proteolysis
negative regulation of mitotic cell cycle
negative regulation of Ras protein signal transduction
negative regulation of membrane protein ectodomain proteolysis
cellular response to organic substance
protease binding
integrin binding
enzyme activator activity
metalloendopeptidase inhibitor activity
metal ion binding
spermatogenesis
central nervous system development
aging
negative regulation of cell population proliferation
response to xenobiotic stimulus
response to hormone
negative regulation of endopeptidase activity
regulation of Rap protein signal transduction
response to cytokine
positive regulation of catalytic activity
negative regulation of catalytic activity
regulation of MAPK cascade
positive regulation of MAPK cascade
regulation of neuron differentiation
positive regulation of neuron differentiation
positive regulation of adenylate cyclase activity
negative regulation of proteolysis
negative regulation of mitotic cell cycle
negative regulation of Ras protein signal transduction
negative regulation of membrane protein ectodomain proteolysis
cellular response to organic substance
protease binding
integrin binding
enzyme activator activity
metalloendopeptidase inhibitor activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1017481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48131 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1017481 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1017481 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1017481 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1017481 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28782 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23071 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100492 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25946 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8589 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6872 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69592 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24844 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17929 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46611 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3967 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23461 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29520 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96071 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78452 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10749490 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749485 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749489 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749491 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749487 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749488 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749486 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10749484 | - | - |
Rat Genome 230 2.0 Array | 1386940_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367823_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367823_at | - | - |
Rat Genome 230 2.0 Array | 1386940_at | - | - |
Affymetrix HT Rat Focus Array | 1367823_at | - | - |
Affymetrix HT Rat Focus Array | 1367823_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386940_at | - | - |
Affymetrix HT Rat Focus Array | 1386940_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000038435.rn.1 | TC1000004206.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000038434.rn.1 | TC1000004206.rn.1 | RTA TC |
Rat Expression Set 230 | 1386940_at | - | - |
Rat Expression Set 230 | 1367823_at | - | - |
Rat Expression Set 230 | 1386940_PM_at | - | - |
Rat Expression Set 230 | 1367823_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10749484 | - | - |
Rat Genome U34 Set | S72594_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21335787 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369398 | - | - |
RatRef-12 Expression BeadChip | ILMN_55617 | - | - |
