Assay Details
Gene Symbol: Rhbdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030034.1 | NP_001025205.1 | 8 - 9 | 1347 | 96 | |
XM_006245977.2 | XP_006246039.1 | 8 - 9 | 5165 | 96 | ||
XM_008767578.1 | XP_008765800.1 | 9 - 10 | 4282 | 96 | ||
XM_008767579.1 | XP_008765801.1 | 9 - 10 | 4157 | 96 | ||
XM_008767580.1 | XP_008765802.1 | 8 - 9 | 2267 | 96 | ||
GenBank mRNA | BC099777.1 | - | 8 - 9 | 1347 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
