Assay Details
Gene Symbol: Slc25a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022398.2 | NP_071793.1 | 2 - 3 | 249 | 123 | |
GenBank mRNA | FQ216657.1 | - | 277 | 123 | ||
U84727.1 | - | 2 - 3 | 249 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
