Assay Details
Gene Symbol: Mpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107036.1 | NP_001100506.1 | 6 - 7 | 1248 | 103 | |
XM_006247095.2 | XP_006247157.1 | 11 - 12 | 1705 | 103 | ||
XM_006247096.2 | XP_006247158.1 | 11 - 12 | 1707 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
