Assay Details
Gene Symbol: Mpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107036.1 | NP_001100506.1 | 7 - 8 | 1423 | 74 | |
XM_006247095.2 | XP_006247157.1 | 12 - 13 | 1880 | 74 | ||
XM_006247096.2 | XP_006247158.1 | 12 - 13 | 1882 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
