Assay Details
Gene Symbol: Trpv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031982.1 | NP_114188.1 | 11 - 12 | 1798 | 104 | |
XM_008767882.1 | XP_008766104.1 | 10 - 11 | 1805 | 104 | ||
GenBank mRNA | AB015231.3 | - | 1049 | 104 | ||
AB040873.1 | - | 10 - 11 | 1718 | 104 | ||
AB041029.1 | - | 9 - 10 | 1538 | 104 | ||
AF029310.1 | - | 11 - 12 | 1798 | 104 | ||
AF158248.2 | - | 5 - 6 | 1131 | 104 | ||
LC008303.1 | - | 6 - 7 | 1038 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
