Assay Details
Gene Symbol: Trpv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031982.1 | NP_114188.1 | 4 - 5 | 685 | 68 | |
XM_008767882.1 | XP_008766104.1 | 3 - 4 | 692 | 68 | ||
GenBank mRNA | AB040873.1 | - | 3 - 4 | 605 | 68 | |
AB041029.1 | - | 3 - 4 | 605 | 68 | ||
AF029310.1 | - | 4 - 5 | 685 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
