Assay Details
Gene Symbol: Trpv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031982.1 | NP_114188.1 | 14 - 15 | 2308 | 69 | |
XM_008767882.1 | XP_008766104.1 | 13 - 14 | 2315 | 69 | ||
GenBank mRNA | AB040873.1 | - | 13 - 14 | 2228 | 69 | |
AF029310.1 | - | 14 - 15 | 2308 | 69 | ||
AF158248.2 | - | 8 - 9 | 1641 | 69 | ||
LC008303.1 | - | 9 - 10 | 1548 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
