Assay Details
Gene Symbol: Rnps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011890.1 | NP_001011890.1 | 7 - 8 | 994 | 92 | |
XM_006245931.2 | XP_006245993.1 | 7 - 8 | 1438 | 92 | ||
XM_006245932.2 | XP_006245994.1 | 8 - 9 | 1137 | 92 | ||
XM_006245933.2 | XP_006245995.1 | 6 - 7 | 933 | 92 | ||
GenBank mRNA | BC079014.1 | - | 7 - 8 | 994 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
