Assay Details
Gene Symbol: Glod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014227.1 | NP_001014249.1 | 2 - 3 | 168 | 82 | |
GenBank mRNA | BC088458.1 | - | 2 - 3 | 168 | 82 | |
FQ230415.1 | - | 2 - 3 | 144 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
