Assay Details
Gene Symbol: Glod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014227.1 | NP_001014249.1 | 4 - 5 | 440 | 86 | |
GenBank mRNA | BC088458.1 | - | 4 - 5 | 440 | 86 | |
FQ210234.1 | - | 4 - 5 | 458 | 86 | ||
FQ230415.1 | - | 4 - 5 | 416 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
