Assay Details
Gene Symbol: Glod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014227.1 | NP_001014249.1 | 1 - 2 | 123 | 67 | |
GenBank mRNA | BC088458.1 | - | 1 - 2 | 123 | 67 | |
FQ210234.1 | - | 1 - 2 | 141 | 67 | ||
FQ230415.1 | - | 1 - 2 | 99 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
