Assay Details
Gene Symbol: Nr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145775.2 | NP_665718.2 | 1 - 2 | 215 | 135 | |
GenBank mRNA | M25804.1 | - | 1 - 2 | 215 | 135 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the M25804.1, NM_145775.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
