Assay Details
Gene Symbol: Nr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113422.1 | NP_001106893.1 | 2 - 3 | 578 | 123 | |
NM_145775.2 | NP_665718.2 | 2 - 3 | 548 | 123 | ||
GenBank mRNA | BC062047.1 | - | 2 - 3 | 578 | 123 | |
M25804.1 | - | 2 - 3 | 548 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
