Assay Details
Gene Symbol: Alox12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105798.1 | NP_001099268.1 | 1 - 2 | 185 | 128 | |
XM_006246659.2 | XP_006246721.1 | 1 - 2 | 227 | 128 | ||
GenBank mRNA | BC168184.1 | - | 1 - 2 | 185 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
