Assay Details
Gene Symbol: Pemt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013003.1 | NP_037135.1 | 4 - 5 | 446 | 82 | |
GenBank mRNA | BC091162.1 | - | 4 - 5 | 486 | 82 | |
L14441.1 | - | 4 - 5 | 446 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
