Assay Details
Gene Symbol: Rara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031528.2 | NP_113716.2 | 4 - 5 | 855 | 88 | |
XM_008767945.1 | XP_008766167.1 | 4 - 5 | 655 | 88 | ||
GenBank mRNA | AJ002940.1 | - | 4 - 5 | 579 | 88 | |
AJ002941.1 | - | 4 - 5 | 595 | 88 | ||
BC099830.1 | - | 4 - 5 | 855 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
