Assay Details
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100158.2 | NP_001093628.1 | 11 - 12 | 1283 | 74 | |
GenBank mRNA | BC105875.1 | - | 11 - 12 | 1228 | 74 | |
FQ234161.1 | - | 10 - 11 | 1005 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
