Assay Details
Gene Symbol: Thoc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024384.2 | NP_077360.2 | 1 - 2 | 276 | 122 | |
GenBank mRNA | BC079149.1 | - | 1 - 2 | 276 | 122 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079149.1, NM_024384.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
