Assay Details
Gene Symbol: Map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030023.1 | NP_001025194.1 | 2 - 3 | 383 | 116 | |
GenBank mRNA | AY880882.1 | - | 2 - 3 | 383 | 116 | |
KC795688.1 | - | 3 - 4 | 416 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
