Assay Details
Gene Symbol: Cdc42ep4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107063.2 | NP_001100533.1 | 1 - 2 | 101 | 110 | |
GenBank mRNA | BC161986.1 | - | 1 - 2 | 101 | 110 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC161986.1, NM_001107063.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
