Assay Details
Gene Symbol: Jmjd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014116.1 | NP_001014138.1 | 6 - 7 | 753 | 84 | |
XM_006245999.2 | XP_006246061.1 | 6 - 7 | 766 | 84 | ||
GenBank mRNA | BC079205.1 | - | 6 - 7 | 753 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
