Assay Details
Gene Symbol: Scrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012142.1 | NP_001012142.1 | 4 - 5 | 608 | 144 | |
XM_006247342.2 | XP_006247404.1 | 4 - 5 | 837 | 144 | ||
XM_008768079.1 | XP_008766301.1 | 4 - 5 | 2121 | 144 | ||
GenBank mRNA | BC078939.1 | - | 4 - 5 | 608 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
