Assay Details
Gene Symbol: Slc16a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247931.2 | XP_006247993.1 | 1 - 2 | 88 | 59 | |
XM_006247934.2 | XP_006247996.1 | 1 - 2 | 88 | 59 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006247931.2, XM_006247934.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
