Assay Details
Gene Symbol: Sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009622.1 | NP_001009622.1 | 4 - 5 | 337 | 86 | |
GenBank mRNA | BC088842.1 | - | 4 - 5 | 337 | 86 | |
FQ215352.1 | - | 2 - 3 | 172 | 86 | ||
FQ215892.1 | - | 2 - 3 | 172 | 86 | ||
FQ216359.1 | - | 2 - 3 | 172 | 86 | ||
FQ216811.1 | - | 2 - 3 | 172 | 86 | ||
FQ217315.1 | - | 2 - 3 | 172 | 86 | ||
FQ217751.1 | - | 4 - 5 | 398 | 86 | ||
FQ219007.1 | - | 2 - 3 | 172 | 86 | ||
FQ219108.1 | - | 2 - 3 | 172 | 86 | ||
FQ219381.1 | - | 4 - 5 | 399 | 86 | ||
FQ220447.1 | - | 2 - 3 | 172 | 86 | ||
FQ224969.1 | - | 2 - 3 | 172 | 86 | ||
FQ227071.1 | - | 2 - 3 | 172 | 86 | ||
FQ233866.1 | - | 2 - 3 | 172 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
