Assay Details
Gene Symbol: Sept6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173429.1 | NP_001166900.1 | 3 - 4 | 393 | 85 | |
XM_006257463.2 | XP_006257525.1 | 3 - 4 | 716 | 85 | ||
XM_008773441.1 | XP_008771663.1 | 3 - 4 | 716 | 85 | ||
XM_008773442.1 | XP_008771664.1 | 3 - 4 | 716 | 85 | ||
XM_008773443.1 | XP_008771665.1 | 3 - 4 | 716 | 85 | ||
XM_008773444.1 | XP_008771666.1 | 3 - 4 | 423 | 85 | ||
GenBank mRNA | BC169051.1 | - | 3 - 4 | 393 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
