Assay Details
Gene Symbol: Muc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012602.1 | NP_036734.1 | 3 - 4 | 1542 | 129 | |
XM_006232590.2 | XP_006232652.1 | 4 - 5 | 1807 | 129 | ||
XM_008761128.1 | XP_008759350.1 | 4 - 5 | 1808 | 129 | ||
GenBank mRNA | BC166505.1 | - | 3 - 4 | 1542 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
