Assay Details
Gene Symbol: Muc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012602.1 | NP_036734.1 | 4 - 5 | 1682 | 80 | |
XM_006232590.2 | XP_006232652.1 | 5 - 6 | 1947 | 80 | ||
XM_008761128.1 | XP_008759350.1 | 5 - 6 | 1948 | 80 | ||
GenBank mRNA | BC166505.1 | - | 4 - 5 | 1682 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
