Assay Details
Gene Symbol: Dera | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749862.3 | XP_003749910.1 | 2 - 3 | 218 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
