Assay Details
Gene Symbol: Dera | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749862.3 | XP_003749910.1 | 4 - 5 | 466 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
