Assay Details
Gene Symbol: Dera | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749862.3 | XP_003749910.1 | 7 - 8 | 843 | 97 | |
GenBank mRNA | FQ212862.1 | - | 2 - 3 | 202 | 97 | |
FQ219539.1 | - | 2 - 3 | 141 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
