Assay Details
Gene Symbol: Atp6v1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198745.2 | NP_942040.1 | 5 - 6 | 474 | 113 | |
GenBank mRNA | BC059155.1 | - | 5 - 6 | 478 | 113 | |
FQ212928.1 | - | 5 - 6 | 113 | |||
FQ212966.1 | - | 5 - 6 | 469 | 113 | ||
FQ213001.1 | - | 5 - 6 | 474 | 113 | ||
FQ213103.1 | - | 5 - 6 | 471 | 113 | ||
FQ213345.1 | - | 5 - 6 | 476 | 113 | ||
FQ213479.1 | - | 5 - 6 | 475 | 113 | ||
FQ213667.1 | - | 5 - 6 | 492 | 113 | ||
FQ213669.1 | - | 5 - 6 | 476 | 113 | ||
FQ214240.1 | - | 5 - 6 | 468 | 113 | ||
FQ214254.1 | - | 5 - 6 | 476 | 113 | ||
FQ214381.1 | - | 5 - 6 | 474 | 113 | ||
FQ214406.1 | - | 5 - 6 | 469 | 113 | ||
FQ214440.1 | - | 5 - 6 | 476 | 113 | ||
FQ214476.1 | - | 5 - 6 | 479 | 113 | ||
FQ214920.1 | - | 5 - 6 | 474 | 113 | ||
FQ216629.1 | - | 5 - 6 | 474 | 113 | ||
FQ224865.1 | - | 5 - 6 | 475 | 113 | ||
FQ230006.1 | - | 5 - 6 | 474 | 113 | ||
FQ233569.1 | - | 5 - 6 | 476 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
