Assay Details
Gene Symbol: Ndufa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100752.1 | NP_001094222.1 | 2 - 3 | 238 | 67 | |
XM_006237500.1 | XP_006237562.1 | 2 - 3 | 321 | 67 | ||
GenBank mRNA | BC091192.1 | - | 2 - 3 | 204 | 67 | |
BC168777.1 | - | 2 - 3 | 238 | 67 | ||
FQ216233.1 | - | 2 - 3 | 238 | 67 | ||
FQ216540.1 | - | 2 - 3 | 238 | 67 | ||
FQ216671.1 | - | 2 - 3 | 228 | 67 | ||
FQ216764.1 | - | 2 - 3 | 228 | 67 | ||
FQ232722.1 | - | 2 - 3 | 236 | 67 | ||
FQ233877.1 | - | 2 - 3 | 306 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
