Assay Details
Gene Symbol: Tuba3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024336.1 | NP_001019507.1 | 3 - 4 | 424 | 100 | |
GenBank mRNA | BC079242.1 | - | 3 - 4 | 424 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
